missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-(2-pyridyl)-L-alanine, 98%, Thermo Scientific Chemicals

Catalog Number 15439488
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
71239-85-5
C13H18N2O4
266.297
MFCD00191190
KMODKKCXWFNEIK-JTQLQIEISA-N
boc-l-2-pyridylalanine, s-2-tert-butoxycarbonyl amino-3-pyridin-2-yl propanoic acid, boc-3-2-pyridyl-l-alanine, boc-l-3-2-pyridyl-alanine, boc-ala 2-pyridyl-oh, boc-2-pal-oh, boc-3-2-pyridyl-ala-oh, boc-2'-pyridyl-l-ala, n-boc-3-2-pyridyl-l-alanine, boc-l-2-pyridylala
2734482
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-2-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CC=N1)C(=O)O
This item is not returnable. View return policy
71239-85-5
C13H18N2O4
266.297
MFCD00191190
KMODKKCXWFNEIK-JTQLQIEISA-N
boc-l-2-pyridylalanine, s-2-tert-butoxycarbonyl amino-3-pyridin-2-yl propanoic acid, boc-3-2-pyridyl-l-alanine, boc-l-3-2-pyridyl-alanine, boc-ala 2-pyridyl-oh, boc-2-pal-oh, boc-3-2-pyridyl-ala-oh, boc-2'-pyridyl-l-ala, n-boc-3-2-pyridyl-l-alanine, boc-l-2-pyridylala
2734482
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-2-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CC=N1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 6416850
Formula Weight 266.29
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Boc-3-(2-pyridyl)-L-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'