Learn More
N-Boc-glycinamide, 95%
Product Code. 15448638
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448638 1 g 1g
15468638 250 mg 250mg
15458638 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15448638 Supplier Thermo Scientific Alfa Aesar Supplier No. H62053.03
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
CAS: 35150-09-5 | C7H14N2O3 | 174.2 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 35150-09-5
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.2
MDL Number MFCD08275106
InChI Key RHONTQZNLFIDCQ-UHFFFAOYSA-N
Synonym boc-glycinamide, tert-butyl 2-amino-2-oxoethyl carbamate, boc-gly-nh2, tert-butyl n-carbamoylmethyl carbamate, n-tert-butoxycarbonyl-l-glycinamide, carbamic acid, 2-amino-2-oxoethyl-, 1,1-dimethylethyl ester, carbamoylmethyl-carbamic acid tert-butyl ester, boc-glycine amide, t-butyl 2-amino-2-oxoethyl carbamate
PubChem CID 10910064
IUPAC Name tert-butyl N-(2-amino-2-oxoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC(=O)N
Specifications
Color White
Linear Formula (CH3)3COCONHCH2CONH2
Quantity 1 g
Formula Weight 174.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-glycinamide