missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-aspartic acid 4-cyclohexyl ester, 95%

Merk:  Alfa Aesar™ H61240.06

Overige gegevens : CAS : 73821-95-1 Gewicht : 0.00500kg

Aantal 5g

 Bekijk meerdere types van dit product via de

Produktcode. 15447748

  • € 29.50 / 5 gram


Geschatte verzenddatum
bij leverancier 22-07-2019
Toevoegen aan winkelmandje

Specificatie

Chemische identificatiekenmerken

73821-95-1
315.366
NLPQIWFEEKQBBN-NSHDSACASA-N
7009416
CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O
C15H25NO6
MFCD00061996
boc-asp ochx-oh, boc-asp ochex-oh, boc-l-aspartic acid 4-cyslohexyl ester, n-boc-l-aspartic acid 4-cyclohexyl ester, 2s-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-asp o-chex, ambotzbaa1074, pubchem12097, 4-cyclohexyl n-boc-l-aspartate, boc-l-aspartic acid b-cyclohexyl ester
(2S)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid

Specificatie

N-Boc-L-aspartic acid 4-cyclohexyl ester
C15H25NO6
3563575
NLPQIWFEEKQBBN-NSHDSACASA-N
(2S)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
7009416
Powder
White
5g
73821-95-1
MFCD00061996
boc-asp ochx-oh, boc-asp ochex-oh, boc-l-aspartic acid 4-cyslohexyl ester, n-boc-l-aspartic acid 4-cyclohexyl ester, 2s-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-asp o-chex, ambotzbaa1074, pubchem12097, 4-cyclohexyl n-boc-l-aspartate, boc-l-aspartic acid b-cyclohexyl ester
CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O
315.366
315.36
95%
93°C to 95°C
Video's
SDS