Learn More
N-Boc-L-felinine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Boc-L-felinine |
| CAS | 879207-98-4 |
| Molecular Formula | C13 H25 N O5 S |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C13H25NO5S/c1-12(2,3)19-11(18)14-9(10(16)17)8-20-13(4,5)6-7-15/h9,15H,6-8H2,1-5H3,(H,14,18)(H,16,17)/t9-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@@H](CSC(C)(C)CCO)C(=O)O |
| IUPAC Name | (2R)-3-(4-hydroxy-2-methylbutan-2-yl)sulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| Molecular Weight (g/mol) | 307.41 |
| Formula Weight | 307.1453 |