missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-Serine Ethyl Ester, TRC

Product Code. 30409974
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409974 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409974 Supplier Toronto Research Chemicals Supplier No. TRCS330055500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-L-Serine Ethyl Ester
CAS 70930-18-6
Molecular Formula C10H19NO5
Quantity 500 mg
Synonym N-(tert-butoxycarbonyl)serine Ethyl Ester, Ethyl (S)-2-[(tert-butoxycarbonyl)amino]-3-hydroxypropanoate, (S)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate
InChI Formula InChI=1S/C10H19NO5/c1-5-15-8(13)7(6-12)11-9(14)16-10(2,3)4/h7,12H,5-6H2,1-4H3,(H,11,14)/t7-/m0/s1
SMILES CCOC([C@@H](NC(OC(C)(C)C)=O)CO)=O
IUPAC Name N-(tert-butoxycarbonyl)serine Ethyl Ester; Ethyl (S)-2-[(tert-butoxycarbonyl)amino]-3-hydroxypropanoate; (S)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate
Molecular Weight (g/mol) 233.26
Formula Weight 233.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.