Learn More
N-Boc-N-methyl-L-serine, 97%
Product Code. p-7054882
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15459948
Brand: Thermo Scientific Alfa Aesar H63314.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 101772-29-6 | C9H17NO5 | 219.237 g/mol
N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101772-29-6
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00037249
InChI Key NJQGIDVCNBZXLG-LURJTMIESA-N
Synonym boc-n-me-ser-oh, boc-n-methyl-l-serine, boc-n-a-methyl-l-serine, boc-n-, a-methyl-l-serine, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, n-boc-n-methyl-l-serine, l-serine,n-1,1-dimethylethoxy carbonyl-n-methyl, boc-meser-oh, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, boc-l-meser-oh
PubChem CID 7009594
IUPAC Name (2S)-3-hydroxy-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
SMILES CC(C)(C)OC(=O)N(C)C(CO)C(=O)O
Specifications
Melting Point 83°C to 87°C
Beilstein 8685334
Quantity 250 mg
Formula Weight 219.23
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-N-methyl-L-serine
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.