missing translation for 'onlineSavingsMsg'
Learn More

N-α-BOC-N-β-Xanthenyl-L-asparagine, 96%, ACROS Organics™

Brand:  Acros Organics 440840050

Additional Details : CAS : 65420-40-8 Weight : 0.05500kg

Quantity 5g

 View more versions of this product

Product Code. 11905511

  • € 128.00 / 5g


Estimated Shipment: 06-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

65420-40-8
412.44
YMGDQLXBNMRJMR-UHFFFAOYNA-N
7017974
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O
C22H24N2O6
MFCD00066157
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid

Specifications

(0.5mmol/mL DMF) clear, colorless
Authentic
1% Max.
170°C to 172°C
65420-40-8
100.0
C22H24N2O6
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O
412.44
412.45
96%
N-α-BOC-N-β-Xanthenyl-L-asparagine
9.5
White
5g
95.0
95% min. (HPLC)
MFCD00066157
YMGDQLXBNMRJMR-UHFFFAOYNA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
7017974
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-alpha-BOC-N-beta-Xanthenyl-L-asparagine

SDS Documents
Certificates