missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-S-tert-butyl-L-cysteine, 98%, Thermo Scientific™

Catalog Number p-7054947
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
56976-06-8
C12H23NO4S
277.379
MFCD00076918
OGARKMDCQCLMCS-QMMMGPOBSA-N
boc-cys tbu-oh, boc-s-tert-butyl-l-cysteine, boc-s-t-butyl-l-cysteine, boc-l-cys tbu-oh, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-1,1-dimethylethyl, 2r-2-tert-butoxycarbonyl amino-3-tert-butylsulfanyl propanoic acid, ambotzbaa1082, boc-s-t-butyl-l-cys, boc-s-tert-butyl-l-cys, n-boc-s-tert-butyl-l-cysteine
11288930
(2R)-3-tert-butylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CSC(C)(C)C)C(=O)O
This item is not returnable. View return policy
56976-06-8
C12H23NO4S
277.379
MFCD00076918
OGARKMDCQCLMCS-QMMMGPOBSA-N
boc-cys tbu-oh, boc-s-tert-butyl-l-cysteine, boc-s-t-butyl-l-cysteine, boc-l-cys tbu-oh, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-1,1-dimethylethyl, 2r-2-tert-butoxycarbonyl amino-3-tert-butylsulfanyl propanoic acid, ambotzbaa1082, boc-s-t-butyl-l-cys, boc-s-tert-butyl-l-cys, n-boc-s-tert-butyl-l-cysteine
11288930
(2R)-3-tert-butylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CSC(C)(C)C)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 277.38
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-Boc-S-tert-butyl-L-cysteine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'