missing translation for 'onlineSavingsMsg'
Learn More

N-Bromoacetamide, 95%

Product Code. p-7043830
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397905 5 g 5g
11307915 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397905 Supplier Thermo Scientific Alfa Aesar Supplier No. L02953.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-15-2 | C2H4BrNO | 137.964 g/mol

Abolishes the rapid inactivation of membrane sodium- and potassium-ion channels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Abolishes the rapid inactivation of membrane sodium- and potassium-ion channels.

Solubility
Soluble in water.

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-15-2
Molecular Formula C2H4BrNO
Molecular Weight (g/mol) 137.964
MDL Number MFCD00037097
InChI Key VBTQNRFWXBXZQR-UHFFFAOYSA-N
Synonym acetamide, n-bromo, unii-0aq6mwh7zb, ccris 4590, 0aq6mwh7zb, acetobromamide, n-bromo-acetamide, sjyhiabiktp@, acmc-209pgg, n-bromoacetamide, powder, lopac-b-2377
PubChem CID 4353
IUPAC Name N-bromoacetamide
SMILES CC(=O)NBr

Specifications

Melting Point 102°C to 106°C
Linear Formula CH3CONHBr
Quantity 5 g
Beilstein 969346
Merck Index 14,1398
Solubility Information Soluble in water.
Formula Weight 137.97
Percent Purity 95%
Chemical Name or Material N-Bromoacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.