missing translation for 'onlineSavingsMsg'
Learn More

N-Bsmoc-glycine, 99%

Product Code. 11305595
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305595 1 g 1g
11325595 250 mg 250mg
11315595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11305595 Supplier Thermo Scientific Alfa Aesar Supplier No. L19737.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 197245-13-9 | C12H11NO6S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197245-13-9
Molecular Formula C12H11NO6S
MDL Number MFCD03792135
Synonym n-bsmoc-glycine, n-benzo b thiophenesulfone-2-methoxycarbonyl glycine, n-benzo b thiophenesulfone-2-methoxycarbonyl aminoacetic acid, 1,1-dioxo-1??-benzothiophen-3-yl methoxy carbonyl amino acetic acid

Specifications

Melting Point 161°C to 163°C
Beilstein 8269937
Quantity 1 g
Formula Weight 297.29
Percent Purity 99%
Chemical Name or Material N-Bsmoc-glycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.