Learn More
n-Butyl propionate, 99%
Product Code. 11470651
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470651 500 g 500g
11460651 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11470651 Supplier Thermo Scientific Alfa Aesar Supplier No. B20125.36
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 590-01-2 | C7H14O2 | 130.187 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 590-01-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009448
InChI Key BTMVHUNTONAYDX-UHFFFAOYSA-N
Synonym butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
PubChem CID 11529
IUPAC Name butyl propanoate
SMILES CCCCOC(=O)CC
Specifications
Density 0.87
Boiling Point 144°C to 146°C
Flash Point 38°C (101°F)
Refractive Index 1.402
Quantity 500 g
UN Number UN3272
Merck Index 14,1587
Formula Weight 130.19
Percent Purity 99%
Chemical Name or Material n-Butyl propionate