Learn More
n-Butyraldehyde-2,2,3,3,4,4,4-d7, CDN
Product Code. 30313276
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313276 0.25 g 0.25g
30318826 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30313276 Supplier CDN Supplier No. CDND59840.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Butanal-2,2,3,3,4,4,4-D7
CAS 951209-46-4
Molecular Formula C4HD7O
Quantity 0.25 g
Synonym 2,2,3,3,4,4,4-d7-Butyraldehyde
InChI Formula InChI=1 S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3,2D2,3D2
SMILES C(C(C([2 H])([2 H])[2 H])([2 H])[2 H])(C=O)([2 H])[2 H]
IUPAC Name butanal-2,2,3,3,4,4,4-d7
Molecular Weight (g/mol) 79.15
Formula Weight 79.1 g/mol
Percent Purity 98 atom % D, min 96% Chemical Purity