Learn More
n-Butyraldehyde-2,2,3,3,4,4,4-d7, CDN
Product Code. 30318826
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318826 0.1 g 0.1g
30313276 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318826 Supplier CDN Supplier No. CDND59840.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Butanal-2,2,3,3,4,4,4-D7
CAS 951209-46-4
Molecular Formula C4HD7O
Quantity 0.1 g
Synonym 2,2,3,3,4,4,4-d7-Butyraldehyde
InChI Formula InChI=1 S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3,2D2,3D2
SMILES C(C(C([2 H])([2 H])[2 H])([2 H])[2 H])(C=O)([2 H])[2 H]
IUPAC Name butanal-2,2,3,3,4,4,4-d7
Molecular Weight (g/mol) 79.15
Formula Weight 79.1 g/mol
Percent Purity 98 atom % D, min 96% Chemical Purity
Show More Show Less
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.