Learn More
n-Butyraldehyde-2,2,3,3,4,4,4-d7, TRC
Specifications
Specifications
| Chemical Name or Material | Butanal-2,2,3,3,4,4,4-D7 |
| CAS | 951209-46-4 |
| Molecular Formula | C4HD7O |
| Quantity | 5 mg |
| Synonym | 2,2,3,3,4,4,4-d7-Butyraldehyde |
| InChI Formula | InChI=1S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3,2D2,3D2 |
| SMILES | C(C(C([2H])([2H])[2H])([2H])[2H])(C=O)([2H])[2H] |
| IUPAC Name | butanal-2,2,3,3,4,4,4-d7 |
| Molecular Weight (g/mol) | 79.15 |
| Formula Weight | 79.1 |