missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cadherin Antibody (CDH2/8862R), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24009C
This item is not returnable.
View return policy
Description
N-Cadherin Monoclonal antibody specifically detects N-Cadherin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| N-Cadherin | |
| Monoclonal | |
| DyLight 650 | |
| ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, N-cadherin 1, Neural cadherin, neural-cadherin | |
| Recombinant fragment (around aa605-905) of human N-Cadherin protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells | |
| 1000 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| CDH2/8862R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction