missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cadherin Antibody (OTI2G7), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72891PE
This item is not returnable.
View return policy
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.
Specifications
| N-Cadherin | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| CDH2 | |
| This N-Cadherin Antibody (OTI2G7) [PE] was developed against a full length recombinant protein of human N-Cadherin (NP_001783) produced in HEK293T cells. | |
| Affinity Purified | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells | |
| 1000.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2G7 | |
| Flow Cytometry | |
| cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
| Mouse | |
| 97.2 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Primate | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction