Learn More
N-Carbobenzyloxy-L-serine b-Lactone, TRC
Specifications
Specifications
| Chemical Name or Material | N-Carbobenzyloxy-L-serine beta-Lactone |
| CAS | 26054-60-4 |
| Molecular Formula | C11 H11 N O4 |
| Quantity | 25 g |
| Synonym | N-Carbobenzyloxy-L-Serine -Ss-Lactone |
| InChI Formula | InChI=1S/C11H11NO4/c13-10-9(7-15-10)12-11(14)16-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,12,14)/t9-/m0/s1 |
| SMILES | O=C(N[C@H]1COC1=O)OCc2ccccc2 |
| IUPAC Name | benzyl N-[(3S)-2-oxooxetan-3-yl]carbamate |
| Molecular Weight (g/mol) | 221.21 |
| Formula Weight | 221.0688 |