Learn More
N-Cbz-D-methionine, TRC
Product Code. 30348794
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348794 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30348794 Supplier Toronto Research Chemicals Supplier No. TRCC22774025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Cbz-D-methionine
CAS 28862-80-8
Molecular Formula C13H17NO4S
Quantity 25 g
Synonym N-[(Phenylmethoxy)carbonyl]-D-methionine
InChI Formula InChI=1S/C13H17NO4S/c1-19-8-7-11(12(15)16)14-13(17)18-9-10-5-3-2-4-6-10/h2-6,11H,7-9H2,1H3,(H,14,17)(H,15,16)/t11-/m1/s1
SMILES O=C(O)[C@H](NC(OCC1=CC=CC=C1)=O)CCSC
IUPAC Name ((benzyloxy)carbonyl)-D-methionine
Molecular Weight (g/mol) 283.34
Formula Weight 283.09