Learn More
N-Cyano-1-chloroformamidine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Cyano-1-chloroformamidine |
| CAS | 25816-30-2 |
| Molecular Formula | C2H2ClN3 |
| Quantity | 2.5 g |
| Synonym | N-Cyano-carbamimidic Chloride, 1-Chloro-N-cyano-formamidine |
| InChI Formula | InChI=1S/C2H2ClN3/c3-2(5)6-1-4/h(H2,5,6) |
| SMILES | NC(=NC#N)Cl |
| IUPAC Name | N'-cyanocarbamimidoyl chloride |
| Molecular Weight (g/mol) | 103.51 |
| Formula Weight | 102.9937 |