missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cyano-3-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanimidamide Maleate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-Cyano-3-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanimidamide Maleate
Specifications
Specifications
| Chemical Name or Material | N-Cyano-3-[((2-[(diaminomethylene)amino]thiazol-4-yl)methyl)sulfanyl]propanimidamide Maleate |
| Analyte or Component Names | N-Cyano-3-[((2-[(diaminomethylene)amino]thiazol-4-yl)methyl)sulfanyl]propanimidamide Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H13 N7 S2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Famotidine Imp. G (EP), N-Cyano-3-[[[2-(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanimidamide Maleate, Famotidine Imp. G (EP) as Maleate, Famotidine Impurity G as Maleate |
| InChI Formula | InChI=1S/C9H13N7S2.C4H4O4/c10-5-14-7(11)1-2-17-3-6-4-18-9(15-6)16-8(12)13;5-3(6)1-2-4(7)8/h4H,1-3H2,(H2,11,14)(H4,12,13,15,16);1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | NC(=Nc1nc(CSCCC(=N)NC#N)cs1)N.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N-cyano-3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanimidamide |
| Molecular Weight (g/mol) | 399.45 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction