missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexylpropyl Deoxynojirimycin, Hydrochloride, TRC

Product Code. 30356786
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30356786 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356786 Supplier Toronto Research Chemicals Supplier No. TRCC9881601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Cyclohexylpropyl Deoxynojirimycin, Hydrochloride
CAS 1221793-31-2
Molecular Formula C15H30ClNO4
Quantity 1 mg
InChI Formula InChI=1S/C15H29NO4.ClH/c17-10-12-14(19)15(20)13(18)9-16(12)8-4-7-11-5-2-1-3-6-11;/h11-15,17-20H,1-10H2;1H/t12-,13+,14-,15-;/m1./s1
SMILES Cl.OC[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)CN1CCCC2CCCCC2
IUPAC Name (2R,3R,4R,5S)-1-(3-cyclohexylpropyl)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
Molecular Weight (g/mol) 323.86
Formula Weight 323.1863
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.