missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Deacetylase/N-Sulfotransferase 2/NDST2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81747F
This item is not returnable.
View return policy
Description
N-Deacetylase/N-Sulfotransferase 2/NDST2 Polyclonal antibody specifically detects N-Deacetylase/N-Sulfotransferase 2/NDST2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunofluorescence
Specifications
| N-Deacetylase/N-Sulfotransferase 2/NDST2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 8509 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunofluorescence | |
| FITC | |
| bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 2, EC 2.8.2.8, Glucosaminyl N-deacetylase/N-sulfotransferase 2, HSST2MGC129697, MGC129705, N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2, NDST-2, N-heparan sulfate sulfotransferase 2, N-HSST 2, NST2 | |
| NDST2 antibody was raised against a 16 amino acid peptide near the amino terminus of human NDST2. The immunogen is located within amino acids 20 - 70 of NDST2. Amino Acid Squence: YVSTSPKAKEPLPLPL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction