Learn More
N-Demethyl Ivabradine Hydrochloride, TRC
Product Code. 30006368
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
50 mg
Unit Size:
2.5mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006368 50 mg 50mg
30004703 5 mg 5mg
30003427 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006368 Supplier Toronto Research Chemicals Supplier No. TRCD29300050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1246638-08-3
Molecular Formula C26 H34 N2 O5 . Cl H
Molecular Weight (g/mol) 491.02
Synonym 2H-3-Benzazepin-2-one, 3-[3-[[[(7S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]amino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-, hydrobromide (1:1)
IUPAC Name 3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methylamino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one;hydrochloride
SMILES Cl.COc1cc2CCN(CCCNC[C@H]3Cc4cc(OC)c(OC)cc34)C(=O)Cc2cc1OC
Specifications
Analyte or Component Names N-Demethyl Ivabradine Hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C26H34N2O5.ClH/c1-30-22-11-17-6-9-28(26(29)14-18(17)12-23(22)31-2)8-5-7-27-16-20-10-19-13-24(32-3)25(33-4)15-21(19)20;/h11-13,15,20,27H,5-10,14,16H2,1-4H3;1H/t20-;/m1./s1
Solution Type Neat
Industry Type Forensics
Quantity 50 mg
Formula Weight 490.2234
Chemical Name or Material N-Demethyl Ivabradine Hydrochloride