missing translation for 'onlineSavingsMsg'
Learn More

N-Demethyl Lincomycin Hydrochloride, TRC

Product Code. 30007212
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007212 2.5 mg 2.5mg
30005959 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007212 Supplier Toronto Research Chemicals Supplier No. TRCD2313202.5MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14600-41-0
Molecular Formula C17 H32 N2 O6 S . Cl H
Molecular Weight (g/mol) 428.97
Synonym D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[[(2S,4R)-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, monohydrochloride (9CI), D-erythro-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-(4-propyl-2-pyrrolidinecarboxamido)-1-thio-, hydrochloride (7CI), D-erythro-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-(4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-, monohydrochloride, trans-α- (8CI), D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[(4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-, monohydrochloride, (2S-trans)-, 4'-Propyl-N-demethyllincomycin hydrochloride, N-Demethyl lincomycin hydrochloride, Methyl 6,8-dideoxy-6-[[[(2S,4R)-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-D-erythro-α-D-galacto-octopyranoside hydrochloride, Lincomycin D hydrochloride, N-Demethyllincomycin hydrochloride, N-Desmethyllincomycin hydrochloride
IUPAC Name (2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES Cl.CCC[C@H]1CN[C@@H](C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C17H32N2O6S.ClH/c1-4-5-9-6-10(18-7-9)16(24)19-11(8(2)20)15-13(22)12(21)14(23)17(25-15)26-3;/h8-15,17-18,20-23H,4-7H2,1-3H3,(H,19,24);1H/t8-,9-,10+,11-,12+,13-,14-,15-,17-;/m1./s1
Formula Weight 428.1748
Percent Purity >95%
Chemical Name or Material N-Demethyl Lincomycin hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.