Learn More
N-Des(5-chloro-2-carvoxythienyl)-N-methylphthalamide Rivaroxaban, TRC
Specifications
Specifications
| Chemical Name or Material | N-Des(5-chloro-2-carvoxythienyl)-N-methylphthalamide Rivaroxaban |
| CAS | 2206360-74-7 |
| Molecular Formula | C23H24N4O6 |
| Quantity | 20 mg |
| Synonym | 1,2-Benzenedicarboxamide, N1-methyl-N2-[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]- (ACI), (S)-N1-Methyl-N2-[[2-oxo-3-[4-(3-oxomorpholino)phenyl]oxazolidin-5-yl]methyl]phthalamide, N1-Methyl-N2-[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]phthalamide |
| InChI Formula | InChI=1S/C23H24N4O6/c1-24-21(29)18-4-2-3-5-19(18)22(30)25-12-17-13-27(23(31)33-17)16-8-6-15(7-9-16)26-10-11-32-14-20(26)28/h2-9,17H,10-14H2,1H3,(H,24,29)(H,25,30)/t17-/m0/s1 |
| SMILES | O=C1N(C[C@H](CNC(=O)C2=C(C(NC)=O)C=CC=C2)O1)C3=CC=C(C=C3)N4C(=O)COCC4 |
| IUPAC Name | (S)-N1-Methyl-N2-((2-oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)phthalamide |
| Molecular Weight (g/mol) | 452.46 |
| Formula Weight | 452.17 |