missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl 8-Chloro-5H-dibenzo[b,e][1,4]diazepine Clozapine, TRC

Product Code. 30361959
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361959 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361959 Supplier Toronto Research Chemicals Supplier No. TRCD2919755MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11,11'-(Piperazine-1,4-diyl)bis(8-chloro-5H-dibenzo[b,e][1,4]-diazepine)
CAS 263366-81-0
Molecular Formula C30 H24 Cl2 N6
Quantity 5 mg
Synonym Quetiapine Dimer Impurity, Quetiapine Bis(dibenzothiazepinyl)piperazine Impurity, 1,4-Bis(dibenzo[b,f][1,4]thiazepin-11-yl)piperazine hydrochloride
InChI Formula InChI=1S/C30H24Cl2N6/c31-19-9-11-25-27(17-19)35-29(21-5-1-3-7-23(21)33-25)37-13-15-38(16-14-37)30-22-6-2-4-8-24(22)34-26-12-10-20(32)18-28(26)36-30/h1-12,17-18,33-34H,13-16H2
SMILES Clc1ccc2Nc3ccccc3C(=Nc2c1)N4CCN(CC4)C5=Nc6cc(Cl)ccc6Nc7ccccc57
IUPAC Name 3-chloro-6-[4-(3-chloro-11H-benzo[b][1,4]benzodiazepin-6-yl)piperazin-1-yl]-11H-benzo[b][1,4]benzodiazepine
Molecular Weight (g/mol) 539.46
Formula Weight 538.144
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.