missing translation for 'onlineSavingsMsg'
Learn More

N-desmethyl Enzalutamide, MedChemExpress

Product Code. 16438168
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438168

Brand: Medchem Express HY70002A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-desmethyl Enzalutamide is the active metabolite of Enzalutamide.N-desmethyl Enzalutamide is the active metabolite of Enzalutamide. N-desmethyl Enzalutamide demonstrates primary and secondary pharmacodynamics of similar potency to Enzalutamide and circulates at approximately the same plasma concentrations as enzalutamide.

N-desmethyl Enzalutamide is the active metabolite of Enzalutamide.N-desmethyl Enzalutamide is the active metabolite of Enzalutamide. N-desmethyl Enzalutamide demonstrates primary and secondary pharmacodynamics of similar potency to Enzalutamide and circulates at approximately the same plasma concentrations as enzalutamide.

Specifications

Chemical Name or Material N-desmethyl Enzalutamide
CAS 1242137-16-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H14F4N4O2S
Quantity 10 mg
Synonym N-desmethyl MDV 3100
Solubility Information DMSO : 100 mg/mL (222.02 mM; Need ultrasonic)
SMILES S=C(N1C2=CC(F)=C(C(N)=O)C=C2)N(C3=CC=C(C(C(F)(F)F)=C3)C#N)C(C(C)1C)=O
Molecular Weight (g/mol) 450.41
Formula Weight 450.41
Percent Purity 96.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.