Learn More
N-Desmethyl Iomeprol Pentaacetate, TRC
Specifications
Specifications
| Chemical Name or Material | N-Desmethyl Iomeprol Pentaacetate |
| CAS | 87785-51-1 |
| Molecular Formula | C26H30I3N3O13 |
| Quantity | 10 mg |
| Synonym | 5-[[2-(Acetyloxy)acetyl]amino]-N1,N3-bis[2,3-bis(acetyloxy)propyl]-2,4,6-triiodo-1,3-benzenedicarboxamide, 5-Acetoxyacetamido-N,N'-bis(2,3-diacetoxypropyl)-2,4,6-triiodoisophthalamide |
| InChI Formula | InChI=1S/C26H30I3N3O13/c1-11(33)41-8-16(44-14(4)36)6-30-25(39)19-21(27)20(23(29)24(22(19)28)32-18(38)10-43-13(3)35)26(40)31-7-17(45-15(5)37)9-42-12(2)34/h16-17H,6-10H2,1-5H3,(H,30,39)(H,31,40)(H,32,38) |
| SMILES | O=C(NCC(OC(=O)C)COC(=O)C)C1=C(I)C(NC(=O)COC(=O)C)=C(I)C(C(=O)NCC(OC(=O)C)COC(=O)C)=C1I |
| IUPAC Name | ((5-(2-acetoxyacetamido)-2,4,6-triiodoisophthaloyl)bis(azanediyl))bis(propane-3,1,2-triyl) tetraacetate |
| Molecular Weight (g/mol) | 973.24 |
| Formula Weight | 972.8913 |