missing translation for 'onlineSavingsMsg'
Learn More

(+)-N-Desmethyl Tramadol, TRC

Product Code. 30012260
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012260 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012260 Supplier Toronto Research Chemicals Supplier No. TRCD2947251MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 147762-57-0
Analyte or Component Names (+)-N-Desmethyl Tramadol
Shipping Condition Room Temperature
Molecular Formula C15H23NO2
Solution Type Neat
SMILES CNC[C@H]1CCCC[C@]1(O)c2cccc(OC)c2
Industry Type Forensics
Molecular Weight (g/mol) 249.35
Quantity 1 mg
Chemical Name or Material (+)-N-Desmethyl Tramadol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.