Learn More
Thermo Scientific Chemicals N(epsilon)-Acetyl-L-lysine, 99%
Click to view available options
Quantity:
1 g
5 g
Description
N(epsilon)-Acetyl-L-lysine is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N(epsilon)-Acetyl-L-lysine is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.
Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml).
Notes
Hygroscopic. Incompatible with oxidizing agents and heat.
N(epsilon)-Acetyl-L-lysine is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.
Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml).
Notes
Hygroscopic. Incompatible with oxidizing agents and heat.
Specifications
Melting Point | 250°C (decomposition) |
Color | White |
Synonym | n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine |
Solubility Information | Soluble in water (miscible),and 80% acetic acid (50mg/ml). |
Formula Weight | 188.2 |
Percent Purity | 99% |
Quantity | 1 g |
Sensitivity | Hygroscopic |
Physical Form | Powder |
Chemical Name or Material | N(epsilon)-Acetyl-L-lysine |