missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-D-alanine pentafluorophenyl ester, 98%

Merk:  Alfa Aesar™ H63713.03

Overige gegevens : CAS : 125043-04-1 Gewicht : 0.00100kg

Aantal 1g

 Bekijk meerdere types van dit product via de

Produktcode. 15440549

  • € 65.40 / 1 gram


Geschatte verzenddatum
bij leverancier 21-06-2019
Toevoegen aan winkelmandje

Specificatie

Chemische identificatiekenmerken

125043-04-1
477.387
CJXZXBGKOSXBFR-LLVKDONJSA-N
16218559
CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C24H16F5NO4
MFCD00153372
fmoc-d-ala-opfp, fmoc-d ala-opfp, fmoc-d-alanine pentafluorophenyl ester, r-perfluorophenyl 2-9h-fluoren-9-yl methoxy carbonyl amino propanoate, fmoc-d-ala-opfp, pentafluorophenyl n-9h-fluoren-9-yl methoxy carbonyl-d-alaninate, d-alanine,n-9h-fluoren-9-ylmethoxy carbonyl-, pentafluorophenyl ester 9ci, d-alanine,n-9h-fluoren-9-ylmethoxy carbonyl-,pentafluorophenyl ester 9ci, 2,3,4,5,6-pentafluorophenyl 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoate
(2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoate

Specificatie

N-Fmoc-D-alanine pentafluorophenyl ester
C24H16F5NO4
fmoc-d-ala-opfp, fmoc-d ala-opfp, fmoc-d-alanine pentafluorophenyl ester, r-perfluorophenyl 2-9h-fluoren-9-yl methoxy carbonyl amino propanoate, fmoc-d-ala-opfp, pentafluorophenyl n-9h-fluoren-9-yl methoxy carbonyl-d-alaninate, d-alanine,n-9h-fluoren-9-ylmethoxy carbonyl-, pentafluorophenyl ester 9ci, d-alanine,n-9h-fluoren-9-ylmethoxy carbonyl-,pentafluorophenyl ester 9ci, 2,3,4,5,6-pentafluorophenyl 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoate
CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
477.387
477.38
98%
125043-04-1
MFCD00153372
CJXZXBGKOSXBFR-LLVKDONJSA-N
(2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoate
16218559
Solid
1g
Video's
SDS