missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-L-methionine sulfone, 98%

Brand:  Alfa Aesar™ H63033.03

Additional Details : CAS : 163437-14-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439478

  • € 53.70 / 1g


In Stock 1
Add to basket

Specifications

Chemical Identifiers

163437-14-7
403.449
KJLKPACOHZKRFM-SFHVURJKSA-N
7016362
CS(=O)(=O)CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C20H21NO6S
MFCD00153361
fmoc-met o2-oh, fmoc-l-methionine sulfone, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfonyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfonylbutanoic acid, ambotzfaa1406, fmoc-met o 2-oh, pubchem22746, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfon, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-4-methylsulfonyl butanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-4-methylsulfonyl butanoic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfonylbutanoic acid

Specifications

N-Fmoc-L-methionine sulfone
C20H21NO6S
fmoc-met o2-oh, fmoc-l-methionine sulfone, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfonyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfonylbutanoic acid, ambotzfaa1406, fmoc-met o 2-oh, pubchem22746, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfon, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-4-methylsulfonyl butanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-4-methylsulfonyl butanoic acid
CS(=O)(=O)CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
403.449
403.45
98%
1g
163437-14-7
MFCD00153361
KJLKPACOHZKRFM-SFHVURJKSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfonylbutanoic acid
7016362
Powder
White
Videos
Documents