missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-proline pentafluorophenyl ester, 98%, Thermo Scientific™

Catalog Number 15400169
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
86060-90-4
C26H18F5NO4
503.425
MFCD00065672
CQBLOHXKGUNWRV-SFHVURJKSA-N
fmoc-pro-opfp, fmoc-l-pro-opfp, fmoc-l-proline pentafluorophenyl ester, 1,2-pyrrolidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl 2-pentafluorophenyl ester, 2s, n-9-fluorenylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluorene-9-ylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluoren-9-ylmethyl 2-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate, 1-o-9h-fluoren-9-ylmethyl 2-o-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate
2733412
1-O-(9H-fluoren-9-ylmethyl) 2-O-(2,3,4,5,6-pentafluorophenyl) (2S)-pyrrolidine-1,2-dicarboxylate
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)OC5=C(C(=C(C(=C5F)F)F)F)F
This item is not returnable. View return policy
86060-90-4
C26H18F5NO4
503.425
MFCD00065672
CQBLOHXKGUNWRV-SFHVURJKSA-N
fmoc-pro-opfp, fmoc-l-pro-opfp, fmoc-l-proline pentafluorophenyl ester, 1,2-pyrrolidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl 2-pentafluorophenyl ester, 2s, n-9-fluorenylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluorene-9-ylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluoren-9-ylmethyl 2-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate, 1-o-9h-fluoren-9-ylmethyl 2-o-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate
2733412
1-O-(9H-fluoren-9-ylmethyl) 2-O-(2,3,4,5,6-pentafluorophenyl) (2S)-pyrrolidine-1,2-dicarboxylate
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)OC5=C(C(=C(C(=C5F)F)F)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 127°C to 129°C
Beilstein 4731096
Formula Weight 503.42
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Fmoc-L-proline pentafluorophenyl ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'