missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-L-isoleucine, 95%, Thermo Scientific Chemicals

Catalog Number 15440439
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
138775-22-1
C22H25NO4
367.45
MFCD00153389
IQIOLCJHRZWOLS-DZLYFNQKNA-N
fmoc-n-me-ile-oh, fmoc-n-methyl-l-isoleucine, fmoc-n-me-lle-oh, n-fmoc-n-methyl-l-isoleucine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-isoleucine, 2s,3s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylpentanoic acid, fmoc-melle-oh, fmoc-l-meile-oh, ambotzfaa1398, pubchem18966
15433034
(2S,3S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylpentanoic acid
CC[C@H](C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
138775-22-1
C22H25NO4
367.45
MFCD00153389
IQIOLCJHRZWOLS-DZLYFNQKNA-N
fmoc-n-me-ile-oh, fmoc-n-methyl-l-isoleucine, fmoc-n-me-lle-oh, n-fmoc-n-methyl-l-isoleucine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-isoleucine, 2s,3s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylpentanoic acid, fmoc-melle-oh, fmoc-l-meile-oh, ambotzfaa1398, pubchem18966
15433034
(2S,3S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylpentanoic acid
CC[C@H](C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 7548444
Formula Weight 367.44
Percent Purity 95%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Fmoc-N-methyl-L-isoleucine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'