Learn More
Alfa Aesar™ N-Fmoc-O-benzyl-N-methyl-L-threonine, 95%
Brand:  Alfa Aesar™ H63833.MD
Additional Details : CAS : 198561-81-8 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15440749
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Chemical Identifiers
198561-81-8
445.515
OJELSPMZRJEODW-CJAUYULYSA-N
16211006
CC(C(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OCC4=CC=CC=C4
C27H27NO5
MFCD00235882
fmoc-methr bzl-oh, fmoc-n-me-thr bzl-oh, fmoc-n-methyl-o-benzyl-l-threonine, fmoc-l-methr bzl-oh, fmoc-nalpha-methyl-o-benzyl-l-threonine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-benzyl-l-threonine, fmoc-o-benzyl-n-methyl-l-threonine, ambotzfaa1554, fmoc-n-alpha-methyl-o-benzyl-l-threonine, fmoc-n-me-thr bzl-oh hplc
(2S,3R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-phenylmethoxybutanoic acid
Specifications
N-Fmoc-O-benzyl-N-methyl-L-threonine
C27H27NO5
fmoc-methr bzl-oh, fmoc-n-me-thr bzl-oh, fmoc-n-methyl-o-benzyl-l-threonine, fmoc-l-methr bzl-oh, fmoc-nalpha-methyl-o-benzyl-l-threonine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-benzyl-l-threonine, fmoc-o-benzyl-n-methyl-l-threonine, ambotzfaa1554, fmoc-n-alpha-methyl-o-benzyl-l-threonine, fmoc-n-me-thr bzl-oh hplc
CC(C(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OCC4=CC=CC=C4
445.515
445.51
95%
198561-81-8
MFCD00235882
OJELSPMZRJEODW-CJAUYULYSA-N
(2S,3R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-phenylmethoxybutanoic acid
16211006
Solid