missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, Environmental Grade, 96+%, Thermo Scientific Chemicals

Catalog Number p-4782013
45.46 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC

For Pesticide Residue, Environmental, and GC Analysis

It is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing.used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Submicron filtered, halogenated residue: 10ng/L (GC-ECD)
TRUSTED_SUSTAINABILITY
Melting Point -90°C
Color Colorless
Appearance Colorless liquid
Density 0.684 g/cm3 at 20°C
Boiling Point 97°C to 99°C
Quantity 1 L
Flash Point -4°C (24°F)
Ignition Point 215°C (419°F)
Odor Petroleum-like
Physical Form Liquid
Refractive Index 1.3877
Assay Percent Range >96%
Beilstein 1730763
Solubility Information Miscible in alcohol,chloroform,and ether. Insoluble in water.
Formula Weight 100.21
Grade Environmental
Chemical Name or Material n-Heptane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'