Learn More
n-Heptane, Environmental Grade, 96+%
Product Code. 11325317
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325317 4 x 1 L 4 L
11335317 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11325317 Supplier Thermo Scientific Alfa Aesar Supplier No. 040978.M1
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
For Pesticide Residue, Environmental, and GC Analysis | CAS: 142-82-5 | C7H16 | 100.21 g/mol
It is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing.used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Submicron filtered, halogenated residue: 10ng/L (GC-ECD)
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC
Specifications
Color Colorless
Appearance Colorless liquid
Quantity 4 x 1 L
Ignition Point 215°C (419°F)
Odor Petroleum-like
Physical Form Liquid
Assay Percent Range >96%
Formula Weight 100.21
Grade Environmental
Chemical Name or Material n-Heptane