missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexyl-d13-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Hexylamine-D13 |
| CAS | 352438-81-4 |
| Molecular Formula | C6 D13 H2 N |
| Quantity | 0.05 g |
| Synonym | 1-Hexan-1,1,2,2,3,3,4,4,5,5,6,6,6-d13-amine (9 CI), 1-Hexanamine-d13, Hexylamine-d13, 1-Aminohexane-1,1,2,2,3,3,4,4,5,5,6,6,6-d13, 1-Hexylamine-D13, Mono-n-hexylamine, n-Hexyl-d13-amine |
| InChI Formula | InChI=1 S/C6H15N/c1-2-3-4-5-6-7/h2-7H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecadeuteriohexan-1-amine |
| Molecular Weight (g/mol) | 114.27 |
| Formula Weight | 114.202 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction