missing translation for 'onlineSavingsMsg'
Learn More

N-Iodosuccinimide, 98%, Thermo Scientific Chemicals

Catalog Number p-4510485
75.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
516-12-1
C4H4INO2
224.99
MFCD00005512
LQZMLBORDGWNPD-UHFFFAOYSA-N
n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
120273
CHEBI:53204
1-iodopyrrolidine-2,5-dione
IN1C(=O)CCC1=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
516-12-1
C4H4INO2
224.99
MFCD00005512
LQZMLBORDGWNPD-UHFFFAOYSA-N
n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
120273
CHEBI:53204
1-iodopyrrolidine-2,5-dione
IN1C(=O)CCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Melting Point 197.0°C to 204.0°C
Color Brown to White-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Quantity 10 g
Merck Index 15,5088
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in acetone,methanol,moderately soluble in dioxane,slightly soluble in methylene chloride and ethyl a,practically insoluble in carbon tetrachloride,eth
Formula Weight 224.99
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Iodosuccinimide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'