Learn More
N-Methyl-1-L-phenylalanine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Methyl-1-L-phenylalanine |
| CAS | 2566-30-5 |
| Molecular Formula | C10H13NO2 |
| Quantity | 10 mg |
| Synonym | (2S)-2-(Methylamino)-3-phenylpropanoate, (S)-2-(Methylamino)-3-phenylpropanoic Acid, (S)-2-Methylamino-3-phenylpropionic Acid, (S)-N-Methylphenylalanine, L-N-Methylphenylalanine, N-Methylphenylalanine, Nalpha-Methyl-L-phenylalanine |
| InChI Formula | InChI=1S/C10H13NO2/c1-11-9(10(12)13)7-8-5-3-2-4-6-8/h2-6,9,11H,7H2,1H3,(H,12,13)/t9-/m0/s1 |
| SMILES | CN[C@@H](Cc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-2-(methylamino)-3-phenylpropanoic acid |
| Molecular Weight (g/mol) | 179.21 |
| Formula Weight | 179.0946 |