missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2,2,2-trifluoroacetamide, Thermo Scientific™

Catalog Number p-7077206
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
815-06-5
C3H4F3NO
127.07
IQNHBUQSOSYAJU-UHFFFAOYSA-N
n-methyltrifluoroacetamide, n-methyl-2,2,2-trifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl, 2,2,2-trifluoro-n-methyl-acetamide, n-metnyl-2,2,2-trifluoroacetamide, methyltrifluoroacetamide, acmc-209plm, n-methyl-trifluoroacetamide, n-methyltrifluoro-acetamide
69948
2,2,2-trifluoro-N-methylacetamide
CNC(=O)C(F)(F)F
This item is not returnable. View return policy
815-06-5
C3H4F3NO
127.07
IQNHBUQSOSYAJU-UHFFFAOYSA-N
n-methyltrifluoroacetamide, n-methyl-2,2,2-trifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl, 2,2,2-trifluoro-n-methyl-acetamide, n-metnyl-2,2,2-trifluoroacetamide, methyltrifluoroacetamide, acmc-209plm, n-methyl-trifluoroacetamide, n-methyltrifluoro-acetamide
69948
2,2,2-trifluoro-N-methylacetamide
CNC(=O)C(F)(F)F
Melting Point 48.0°C to 52.0°C
Boiling Point 156.0°C to 157.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CF3CONHCH3
Quantity 5 g
Formula Weight 127.07
Percent Purity ≥97.5% (GC)
Chemical Name or Material N-Methyl-2,2,2-trifluoroacetamide
compliance-icons
Product Identifier
  • N-Methyl-2,2,2-trifluoroacetamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'