Learn More
N-Methyl-dl-valine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N-Methyl-dl-valine Hydrochloride |
| CAS | 2566-32-7 |
| Molecular Formula | C6 H13 N O2 |
| Quantity | 1 g |
| Synonym | N-Methyl-dl-valine, HCl |
| InChI Formula | InChI=1S/C6H13NO2/c1-4(2)5(7-3)6(8)9/h4-5,7H,1-3H3,(H,8,9) |
| SMILES | CNC(C(C)C)C(=O)O |
| IUPAC Name | 3-methyl-2-(methylamino)butanoic acid |
| Molecular Weight (g/mol) | 131.173 |
| Formula Weight | 131.095 |