missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-L-proline, 98%, Thermo Scientific Chemicals

Catalog Number p-7054821
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
475-11-6
C6H11NO2
129.159
MFCD00011565
CWLQUGTUXBXTLF-YFKPBYRVSA-N
n-methyl-l-proline, h-n-me-pro-oh, 1-methylproline, 1-methyl-l-proline, 2s-1-methylpyrrolidine-2-carboxylic acid, s-1-methylpyrrolidine-2-carboxylic acid, l-proline, 1-methyl, n-methyl proline, proline, 1-methyl, s-1-methyl-pyrrolidine-2-carboxylic acid
643474
(2S)-1-methylpyrrolidine-2-carboxylic acid
CN1CCCC1C(=O)O
This item is not returnable. View return policy
475-11-6
C6H11NO2
129.159
MFCD00011565
CWLQUGTUXBXTLF-YFKPBYRVSA-N
n-methyl-l-proline, h-n-me-pro-oh, 1-methylproline, 1-methyl-l-proline, 2s-1-methylpyrrolidine-2-carboxylic acid, s-1-methylpyrrolidine-2-carboxylic acid, l-proline, 1-methyl, n-methyl proline, proline, 1-methyl, s-1-methyl-pyrrolidine-2-carboxylic acid
643474
(2S)-1-methylpyrrolidine-2-carboxylic acid
CN1CCCC1C(=O)O

N-Methyl-L-proline, is an aminoacid which is widely used in pharmaceuticals and food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyl-L-proline, is an aminoacid which is widely used in pharmaceuticals and food industry.

Solubility
Soluble in water and methanol(both 50 mg/ml-clear-colorless solution)

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Refractive Index 1.5
Solubility Information Soluble in water and methanol(both 50mg/ml-clear-colorless solution)
Formula Weight 129.16
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material N-Methyl-L-proline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'