Learn More
N-Methylform-d1-amide, CDN
Product Code. 30315578
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315578 1 g 1g
30311510 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315578 Supplier CDN Supplier No. CDND54201G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methylformamide-1-D
CAS 26103-38-8
Molecular Formula C2H4DNO
Quantity 1 g
Synonym Formamide-1-d, N-methyl- (7 CI, 8 CI, 9 CI), N-Methylformamide-D, N-Methylformamide-1-D, N-Methylformamide-d
InChI Formula InChI=1 S/C2H5NO/c1-3-2-4/h2H,1H3,(H,3,4)/i2D
SMILES [2 H]C(NC)=O
IUPAC Name N-methylformamide-1-d
Molecular Weight (g/mol) 60.07
Formula Weight 60.04 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity