Learn More
N-Methylfurfurylamine, 95%
Product Code. 10235194 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. 10235194
Brand: Thermo Scientific Acros 397270050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 4753-75-7 | C6H9NO | 111.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4753-75-7
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00020961
InChI Key DGLIOWSKNOCHEX-UHFFFAOYSA-N
Synonym n-methylfurfurylamine, 1-furan-2-yl-n-methylmethanamine, 2-furanmethanamine, n-methyl, furfurylamine, n-methyl, 2-furylmethyl methylamine, n-furfuryl-n-methylamine, methylfurfurylamine, furan-2-ylmethyl methyl amine, 2-furyl-n-methylmethanamine, furan-2-yl methyl methyl amine
PubChem CID 78492
IUPAC Name 1-(furan-2-yl)-N-methylmethanamine
SMILES CNCC1=CC=CO1
Specifications
Density 0.9820g/mL
Boiling Point 148.0°C to 152.0°C
Flash Point 40.6°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.982
Formula Weight 111.14
Percent Purity 95%
Chemical Name or Material N-Methylfurfurylamine