missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Myc Antibody (NMYC-1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-109F
This item is not returnable.
View return policy
Description
n-Myc Monoclonal specifically detects n-Myc in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| n-Myc | |
| Monoclonal | |
| FITC | |
| P04198 | |
| MYCN | |
| A synthetic peptide (SPYVESEDAPPQKC) conjugated to KLH, corresponding to amino acids 327-339 of human n-Myc. [UniProt# P04198] | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 4613 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| NMYC-1 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| BHLHE37, bHLHe37N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, MODED, neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene, N-myc, NMYCneuroblastoma MYC oncogene, ODED, oncogene NMYC, pp65/67, v-myc avian myelocytomatosis viral related oncogene, neuroblastoma derived, v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody does not react with c-Myc or L-Myc. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction