missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N myc interactor Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-763IR
This item is not returnable.
View return policy
Description
N myc interactor Polyclonal antibody specifically detects N myc interactor in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| N myc interactor | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| Nmi, N-myc (and STAT) interactor, N-myc and STAT interactor, N-myc interactor, N-myc-interactor | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human N-myc and STAT interactor using the numbering given in entry NP_004679.1 (GeneID 9111). | |
| 0.1 mL | |
| Cancer, Signal Transduction, Tumor Suppressors | |
| 9111 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction