Learn More
Alfa Aesar™ N-n-Butyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%
Brand:  Alfa Aesar™ H65917.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15440007
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Chemical Identifiers
MFCD27976901
Specifications
N-n-Butyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide
MFCD27976901
Solid
1g
231.34
95%