missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diboc 1-O-t-Butyl 5-Deoxo L-Glutamic Acid, TRC

Product Code. 30379013
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379013 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379013 Supplier Toronto Research Chemicals Supplier No. TRCD41950010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Diboc 1-O-t-Butyl 5-Deoxo L-Glutamic Acid
CAS 257883-71-9
Molecular Formula C19H35NO7
Quantity 10 mg
Synonym N,N-Bis[(1,1-dimethylethoxy)carbonyl]-L-glutamic Acid 1-(1,1-Dimethylethyl) Ester
InChI Formula InChI=1S/C19H35NO7/c1-17(2,3)25-14(22)13(11-10-12-21)20(15(23)26-18(4,5)6)16(24)27-19(7,8)9/h13,21H,10-12H2,1-9H3/t13-/m0/s1
SMILES O=C(OC(C)(C)C)[C@@H](N(C(OC(C)(C)C)=O)C(OC(C)(C)C)=O)CCCCO
IUPAC Name tert-butyl (2S)-2-[bis[(2-methylpropan-2-yl)oxycarbonyl]amino]-5-hydroxypentanoate
Molecular Weight (g/mol) 389.48
Formula Weight 389.2414
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.