Learn More
N,N-Diethylnicotinamide (Nikethamide), TRC
Product Code. 30040403
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040403 2.5 g 2.5g
30138018 25 g 25g
30136166 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30040403 Supplier Toronto Research Chemicals Supplier No. TRCN4600002.5G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59-26-7
Molecular Formula C10 H14 N2 O
Molecular Weight (g/mol) 178.23
Synonym 3-Pyridinecarboxamide, N,N-diethyl-, Nicotinamide, N,N-diethyl- (8CI), N,N-Diethyl-3-pyridinecarboxamide, 3-(N,N-Diethylcarbamoyl)pyridine, Anacardone, Anacordone, Astrocar, Canfodiamina, Carbamidal, Cardamine, Cardiamid, Cardiamide, Cardiamine, Cardimon, Coracon, Coractiv N, Coramine, Coramine (pharmaceutical), Cordiamin, Cordiamine, Corediol, Cormed, Cormid, Cormotyl, Cornotone, Corvin, Corvitol, Corvotone, Diethylnicotinamide, Dynacoryl, Ecoran, Eucoran, Kordiamin, N,N-Diethylnicotinamide, NSC 130820, NSC 169863, NSC 62103, Ni-Cor, Niamine, Nicamide, Nicetamide, Nicethamide, Nicorine, Nicotinic acid diethylamide, Nikardin, Niketamide, Niketamine, Nikethamide, Niketharol, Nikethyl, Nikorin, Niquetamide, Percoral, Pyricardyl, Pyridine-3-carboxydiethylamide, Pyridine-3-carboxylic acid diethylamide, Reformin, Rehormin, Salvacard, Sancora, Solyacord, Stimulin, Ventramine
IUPAC Name N,N-diethylpyridine-3-carboxamide
SMILES CCN(CC)C(=O)c1cccnc1
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C10H14N2O/c1-3-12(4-2)10(13)9-6-5-7-11-8-9/h5-8H,3-4H2,1-2H3
Formula Weight 178.1106
Chemical Name or Material Nikethamide